Learn More
R&D Systems™ Recombinant Mouse TSLPR Fc Chimera Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 57914 |
| Molecular Weight (g/mol) | 49.3 kDa (monomer) |
| Name | TSLPR/CRLF2 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse TSLP R protein, (N-terminus) Mouse TSLP R (Ala20-Leu233) Accession &Num; BAA92159 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | CRL2, CRL2cytokine receptor CRL2 precusor, CRLF2, CRLF2Y, CRLM-2, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, PCOR1, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP R, TSLP receptor, TSLPR |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.