Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse USAG1 Protein
SDP

Catalog No. 9008SD050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-08S-D050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-08S-D050 Supplier R&D Systems™ Supplier No. 9008SD050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse USAG1 Protein is derived from E. coli. The Recombinant Mouse USAG1 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9CQN4
For Use With (Application) Bioactivity
Gene ID (Entrez) 25928
Molecular Weight (g/mol) Observed M.W.: 21kDa, reducing conditions, Theoretical M.W.: 21kDa
Quantity 50 μg
Source E. coli-derived mouse USAG1 protein Phe24-Ser206,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CDA019, cystine-knot containing secreted protein, DKFZp564D206, Ectodermal BMP inhibitor, ECTODIN, sclerostin domain containing 1, sclerostin domain-containing protein 1, SOSTDC1, USAG-1, USAG1uterine sensitization-associated protein-1, Uterine sensitization-associated gene 1 protein, WISE
Common Name USAG1/SOSTDC1
Gene Symbol Sostdc1
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human USAG1 is immobilized at 1μg/mL (100μL/well), the concentration of Recombinant Mouse LRP-6 that produces 50% of the optimal binding response is approximately 0.1-0.6μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System E. coli-derived Phe24-Ser206, with an N-terminal Met
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.