Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Wnt-1/sFRP-1 Complex Protein

Catalog No. 9765WN010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
97-65W-N010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 97-65W-N010 Supplier R&D Systems™ Supplier No. 9765WN010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Wnt-1/sFRP-1 Complex Protein is derived from CHO. The Recombinant Mouse Wnt-1/sFRP-1 Complex Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P04426, Q8C4U3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, Trehalose and with BSA as a carrier protein.
Gene ID (Entrez) 7471
Molecular Weight (g/mol) 38 kDa (Wnt-1) and 33 kDa (sFRP-1)
Name FrzA, Int-1, INT1Proto-oncogene Int-1 homolog, proto-oncogene Wnt-1, SARP-2, sFRP1, wingless-type MMTV integration site family, member 1, wingless-type MMTV integration site family, member 1 (oncogene INT1), Wnt1
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-1 protein, (N-terminus) Mouse Wnt-1 (Ala28-Leu370) Accession &Num; P04426 Mouse sFRP-1 (Ser32-Lys314) Accession &Num; Q8C4U3 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 100μg/mL in PBS.
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.