Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Wnt-3a, Biotinylated Protein

Catalog No. BT1324010CF
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT1324010CF 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT1324010CF Supplier R&D Systems™ Supplier No. BT1324010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Wnt-3a, Biotinylated Protein is derived from CHO. The Recombinant Mouse Wnt-3a, Biotinylated Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P27467
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Gene ID (Entrez) 89780
Molecular Weight (g/mol) 37 kDa (unlabeled)
Name MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A, Wnt3a
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-3a protein Ser19-Lys352
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Conjugate Biotin
Species Mouse
Reconstitution Reconstitute at 100μg/mL in PBS.
Purity or Quality Grade >75%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.