Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Wnt-8a Protein

Catalog No. 8419WN010CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS with BSA as a carrier protein
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
84-19W-N010CF Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS
84-19W-N010 Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS with BSA as a carrier protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 84-19W-N010CF Supplier R&D Systems™ Supplier No. 8419WN010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Wnt-8a Protein is derived from CHO. The Recombinant Mouse Wnt-8a Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_033316
Formulation Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS
Gene ID (Entrez) 20890
Molecular Weight (g/mol) 37kDa
Name Wnt-8a
Preparation Method Reconstitute at 100μg/mL in PBS.
Quality Control Testing SDS-PAGE: 40-62kDa, reducing conditions
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-8a protein Ala20-Trp354
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ala20 & Trp23
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.