Learn More
Novus Biologicals™ Recombinant Mycobacterium tuberculosis Ab85A His Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing, 10% glycerol |
| Molecular Weight (g/mol) | 32.8kDa |
| Name | TB Ab85A Protein |
| Purification Method | SDS-PAGE |
| Quantity | 50 μg |
| Immunogen | Antigen 85A, 43-338 aa, Mycobacterium tuberculosis, His tag, Baculovirus (NP_218321.1). |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Purity or Quality Grade | >90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.