Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant N/A Lewis X
SDP

Catalog No. GL306100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 pmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GL306100 100 pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GL306100 Supplier R&D Systems™ Supplier No. GL306100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Specifications

Content And Storage Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Quantity 100 pmol
Target Entry Lewis X
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.