Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant P. heparinus Heparinase II Protein

Catalog No. 6336GH010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6336GH010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 6336GH010 Supplier R&D Systems™ Supplier No. 6336GH010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to liberate oligosaccharides from heparin. The specific activity is >750 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number YP_003092674
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8254931
Molecular Weight (g/mol) M.W. (Observed): 66-75 kDa, reducing conditions; M.W. (theoretical): 86 kDa
Name Heparinase II
Quantity 10 μg
Source E. coli-derived p. heparinus Heparinase II protein Ala26-Arg772, with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.)
Protein Heparinase II
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.