Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant P. multocida Hyaluronan Synthase/HAS Protein

Catalog No. 9585GT050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
95-85G-T050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 95-85G-T050 Supplier R&D Systems™ Supplier No. 9585GT050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant P. multocida Hyaluronan Synthase/HAS Protein is derived from E. coli. The Recombinant P. multocida Hyaluronan Synthase/HAS Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number Q7BLV3
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2μm filtered solution in Tris and NaCl.
Molecular Weight (g/mol) 81 kDa
Quantity 50 μg
Source E. coli-derived p. multocida Hyaluronan Synthase protein Met1-Ile703,with C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species P. multocida
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.