Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Parasite HpARI His Protein
SDP

Catalog No. NB171726 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171726 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171726 Supplier Novus Biologicals™ Supplier No. NBP318165
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Binds to human and mouse IL-33

Specifications

Concentration LYOPH
For Use With (Application) Bioactivity, ELISA
Formulation Lyophilized from PBS
Molecular Weight (g/mol) M.W. Theoretical: 40 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.01 EU/μg purified protein (LAL test; Lonza).
Gene Alias H. polygyrus Alarmin Release Inhibitor
Reconstitution Reconstitute with 50 ul sterile water (After reconstitution: 1mg/ml)
Research Category Immunology
Purity or Quality Grade >90% SDS-PAGE
Protein HpARI
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.