Learn More
R&D Systems™ Recombinant Plant Arachis hypogaea 8.0101 Strep (N-Term) Protein
Shop All Novus Products

Specifications
Specifications
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | Lyophilized from a 0.22 um filtered solution in Tris buffer (100 mM Tris, 150 mM NaCl, pH 8) |
| Molecular Weight (g/mol) | M.W. Theoretical: 18.3 kDa |
| Quantity | 0.1 mg |
| Storage Requirements | Store at -20 to -70C. Avoid freeze-thaw cycles. |
| Gene Alias | rAra h 8 |
| Reconstitution | Reconstitute with sterile deionized water with or without azide. |
| Product Line | Novus |
| Purity or Quality Grade | >95%, by HPLC |
| Protein | Arachis hypogaea 8.0101 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.