Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Plant Betula verrucosa 4.0101 Strep (N-Term) Protein
SDP

Catalog No. NB183349 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183349 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183349 Supplier Novus Biologicals™ Supplier No. NBP306723
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

For Use With (Application) ELISA, Flow Cytometry, SDS-PAGE
Formulation Lyophilized from a 0.22 um filtered solution in Tris buffer (100 mM Tris, 150 mM NaCl, pH 8)
Molecular Weight (g/mol) M.W. Theoretical: 10.8 kDa
Quantity 0.1 mg
Storage Requirements Store at -20 to -70C. Avoid freeze-thaw cycles.
Gene Alias rBet v 4
Reconstitution Reconstitute with sterile deionized water with or without azide.
Purity or Quality Grade >95%, by HPLC
Protein Betula verrucosa 4.0101
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.