Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Porcine IL-3 Protein
SDP

Catalog No. NB397978 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397978 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397978 Supplier R&D Systems™ Supplier No. NBP311061

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Recombinant Protein

Specifications

For Use With (Application) Western Blot, ELISA
Formulation PBS
Gene ID (Entrez) 3562
Molecular Weight (g/mol) MolecularWeight-theroretical: 13.7 kDa
Quantity 5 μg
Storage Requirements Store at -20°C.
Gene Alias Hematopoietic growth factor, IL-3 mgC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Gene Symbol IL3
Conjugate Unconjugated
Species Porcine
Recombinant Recombinant
Reconstitution Reconstitute with 20 uL sterile PBS containing 0.1% carrier protein. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. We recommend that the vial is gently mixed after reconstitution.)
Form Purified
Research Category Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE
Protein IL-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.