Learn More
Active Motif Recombinant PPARγ protein
SDP
Supplier:  Active Motif 81214
Encompass_Preferred
PPARg (Peroxisome Proliferator Activated Receptor Gamma), also called as PPARG or NR1C3, is a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. PPAR-gamma is a nuclear receptor that binds peroxisome proliferators such as hypolipidemic drugs and fatty acids. Once activated by a ligand, it can bind to DNA specific PPAR response elements (PPRE) and modulate the transcription of its target genes, such as acyl-CoA oxidase. It therefore controls the peroxisomal beta-oxidation pathway of fatty acids. It's also a key regulator of adipocyte differentiation and glucose homeostasis. […] For more information, please visit: https://www.activemotif.com/catalog/details/81214.
Catalog No. NC1852180
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.