Learn More
R&D Systems™ Recombinant Pro-TNF-alpha Fusion Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Urea, NaCl, NaH 2 PO 4 and DTT. |
| Gene ID (Entrez) | 7124 |
| Molecular Weight (g/mol) | 45 kDa |
| Name | Pro TNF-alpha Fusion Protein |
| Quantity | 10 μg |
| Source | E. coli-derived human TNF-alpha protein, (N-terminus) Bacterial protein, (N-terminus) Fusion Partner Human Pro-TNF-alpha (Gly57-Ala76) Accession &Num; P01375 Human Mature TNF-alpha (Val77-Leu233) Accession &Num; P01375 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.