Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein CTD, His Tag (MALS-verified)
Supplier:  ACROBiosystems S1DC52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 S1 protein CTD, His Tag (MALS-verified) (S1D-C52H3) is expressed from human 293 cells (HEK293). It contains AA Asn 334 - Pro 527 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.6 kDa. The protein migrates as 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 1 μg/mL (100 μL/well) can bind SARS-CoV-2 S1 protein CTD, His Tag (MALS-verified) (Cat. No. S1D-C52H3) with a linear range of 0.6-20 ng/mL (QC tested).; Immobilized SARS-CoV-2 S1 protein CTD, His Tag (MALS-verified) (Cat. No. S1D-C52H3) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) with a linear range of 0.1-3 ng/mL (Routinely tested).
Catalog No. 50-201-9604
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.