Learn More
ACROBiosystems MERS S protein RBD, His Tag
Supplier:  ACROBiosystems SPDM52H61MG
Encompass_Preferred
Recombinant Protein;1MG;MERS S protein RBD, His Tag (SPD-M52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 367 - Tyr 606 (Accession # K0BRG7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.1 kDa.
Catalog No. 50-201-9462