Learn More
ACROBiosystems Human IL-7-P2A Protein, Tag Free
Supplier:  ACROBiosystems ILAH521850UG
Encompass_Preferred
Recombinant Protein;50UG;Human IL-7-P2A, Tag Free (ILA-H5218) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession # P13232-1).;This protein carries no "tag". The protein has a calculated MW of 19.6 kDa. The protein migrates as 26-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-7-P2A, Tag Free (Cat. No. ILA-H5218) at 2 μg/mL (100 μL/well) can bind Human IL-7 R alpha, Fc Tag (Cat. No. ILA-H5258) with a linear range of 0.6-39 ng/mL (QC tested).
Catalog No. 50-201-9773
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.