Learn More
R&D Systems™ Recombinant Protein A His (C-Term) Cys (C-Term) Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from additive free solution. |
| Gene ID (Entrez) | 3919448 |
| Molecular Weight (g/mol) | 34.8 kDa |
| Quantity | 10 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of Protein A as determined by LAL method. |
| Gene Alias | Immunoglobulin G binding protein A, SPA, Staphylococcal protein A |
| Reconstitution | Reconstitute with distilled water or saline. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.