Learn More
Fina Biosolutions LLC CRM197 Carrier Protein | 5mg
Supplier:  Fina Biosolutions LLC CRM1975MG
Encompass
CRM197, a genetically detoxified diphtheria toxin, is widely used as a carrier protein in conjugate vaccines, including effective conjugate vaccines against Streptococcus pneumoniae, Haemophilus influenzae b and Neisseria meningitidis. Fina BioSolutions has developed a highly efficient E. coli expression system for CRM197. We express soluble, properly folded CRM197 in the cytoplasm of E.coli. High expression levels and a simple purification scheme allow us to produce CRM197 at low cost and price it accordingly.
Catalog No. NC1716984
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.