Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant R. sanguineus Evasin-3 His-tag Protein

Catalog No. 10034EV050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-034-EV050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-034-EV050 Supplier R&D Systems™ Supplier No. 10034EV050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant R. sanguineus Evasin-3 His-tag Protein is derived from Sf 21 (baculovirus). The Recombinant R. sanguineus Evasin-3 His-tag Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P0C8E8
For Use With (Application) Bioactivity
Molecular Weight (g/mol) 7.8 kDa
Quantity 50 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived r. sanguineus Evasin-3 protein Leu21-Arg86. with a C-terminal 6-His tag
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity Evasin-3
Conjugate Unconjugated
Species R. sanguineus
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.