Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat 4-1BB/TNFRSF9 Fc Chimera Protein

Catalog No. 79684B050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79684B050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 79684B050 Supplier R&D Systems™ Supplier No. 79684B050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Rat 4-1BB/TNFRSF9/CD137 Fc Chimera is present at 100 ng/mL, the concentration of Recombinant Mouse 4-1BB Ligand/TNFSF9 (Catalog # 1246-4L ) that produces 50% of the optimal binding response is approximately 5-25 ng/mL.

Specifications

Accession Number NP_001020944
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3604
Molecular Weight (g/mol) M.W. (Observed): 60-65 kDa, reducing conditions; M.W. (theoretical): 44.9 kDa (monomer)
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived rat 4-1 BB/TNFRSF9/CD137 protein Rat 4-1 BB (Thr24-Val188) (N-terminus) Accession # NP_001020944 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol Tnfrsf9
Product Type 4-1BB/TNFRSF9/CD137 Fc Chimera
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein 4-1 BB/TNFRSF9/CD137
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.