Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat C-Reactive Protein/CRP Protein
SDP

Catalog No. 1744CRB200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-44C-RB200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-44C-RB200 Supplier R&D Systems™ Supplier No. 1744CRB200
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat C-Reactive Protein/CRP Protein is derived from NS0. The Recombinant Rat C-Reactive Protein/CRP Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P48199
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in Tris, NaCl and CaCl2.
Gene ID (Entrez) 1401
Molecular Weight (g/mol) 25 to 38 kDa
Quantity 200 μg
Source Mouse myeloma cell line,NS0-derived rat C-Reactive Protein/CRP protein His20-Ser230,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name C-Reactive Protein; C-reactive protein, pentraxin-related; CRP; MGC88244; pentraxin 1; PTX1MGC149895
Biological Activity 0.5 to 3μg/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.