Learn More
R&D Systems™ Recombinant Rat Cathepsin C/DPPI Protein
Description
Order Info
The product is shipped with polar packs. Upon receipt, store it immediately
Specifications
Specifications
| Accession Number | P80067 |
| Formulation | Supplied as a 0.2μm filtered solution in MES, NaCl. |
| Molecular Weight (g/mol) | 51 kDa |
| Name | Cathepsin C/DPPI, Carrier Free |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived rat Cathepsin C/DPPI protein Asp25-Leu462,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.