Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat CD40/TNFRSF5 Fc Chimera Protein
SDP

Catalog No. 8229CD050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
82-29C-D050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 82-29C-D050 Supplier R&D Systems™ Supplier No. 8229CD050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to inhibit CD40 Ligand-induced proliferation of mouse B cells. The ED50 for this effect is 0.1-0.6 μg/mL.

Specifications

Accession Number Q4QQW2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and Trehalose.
Gene ID (Entrez) 958
Molecular Weight (g/mol) M.W. (Observed): 57-63 kDa, reducing conditions; M.W. (theoretical): 46 kDa (monomer)
Name CD40/TNFRSF5 Fc Chimera
Preparation Method Reconstitute at 200μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 57-63kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived rat CD40/TNFRSF5 protein Rat CD40 (Val24-Arg193) (N-terminus) Accession # Q4QQW2 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Val24
Without Additives Carrier Protein
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Cd40
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Protein CD40/TNFRSF5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.