Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat CLEC10A/CD301 Protein
SDP

Catalog No. 9037CL050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-37C-L050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-37C-L050 Supplier R&D Systems™ Supplier No. 9037CL050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat CLEC10A/CD301 Protein is derived from CHO. The Recombinant Rat CLEC10A/CD301 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_071788
For Use With (Application) Bioactivity
Gene ID (Entrez) 10462
Molecular Weight (g/mol) Observed M.W.: 34-42kDa, reducing conditions, Theoretical M.W.: 29kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived rat CLEC10A/CD301 protein Gln59-Ser306,with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CD301, CD301 antigen, CD301C-type lectin domain family 10 member A, CLECSF13, CLECSF14, CLECSF14macrophage C-type lectin, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 14 (macrophage-derived), C-type lectin domain family 10, member A, C-type lectin superfamily member 14, HML2, HMLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 13 (macrophage-derived), Macrophage lectin 2, macrophage lectin 2 (calcium dependent), MGL
Common Name CLEC10A/CD301
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Rat CLEC10A/CD301 is immobilized at 2.5μg/mL (100μL/well), the concentration of biotinylated Recombinant Viral EBOV GP that produces 50% of the optimal binding response is approximately 1-6μg/mL
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Gln59-Ser306, with an N-terminal 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.