Learn More
Novus Biologicals™ Recombinant Rat CXCL2/GRO beta/MIP-2/CINC-3 Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Accession Number | P30348, P30348, P30348 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 2920 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 7.9 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of CXCL2/GRO beta/MIP-2/CINC-3 AS determined by LAL method. |
| Gene Alias | chemokine (C-X-C motif) ligand 2, CINC-2 A, CINC-3/CXCL2, C-X-C Motif Chemokine 2, GRO2, GRO2 oncogene, GROB, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP2, MIP-2 A, MIP2A, MIP2-alpha, SCYB2 |
| Gene Symbol | CXCL2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.