Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat FABP1/L-FABP Protein

Catalog No. 9468CL100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9468CL100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9468CL100 Supplier R&D Systems™ Supplier No. 9468CL100
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant Rat FABP1/L-FABP Protein CF is a small cytoplasmic lipid binding protein that is expressed in a tissue specific manner and is involved in intracellular lipid transport.

The Recombinant Rat FABP1/L-FABP Protein is derived from E. coli.

Specifications

Accession Number P02692
Characterization Bioactivity not tested
For Use With (Application) Flow Cytometry
Formulation Supplied as a 0.2μm filtered solution in PBS, DTT and Glycerol.
Gene ID (Entrez) 2168
Molecular Weight (g/mol) 15kDa
Quantity 100 μg
Source E. coli-derived rat FABP1/L-FABP protein, (N-terminus) Met HHHHHH GS Rat FABP1/L-FABP (Met1-Ile127) Accession &Num; P02692 (C-terminus)
Storage Requirements 6 months from date of receipt, 2 to 8° C as supplied. 3 months, 2 to 8° C under sterile conditions after opening.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, LFABP, L-FABP, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein
Product Type Recombinant Protein
Conjugate Unconjugated
Species Rat
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.