Learn More
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | prior to lyophilization: phosphate buffered saline (PBS); 125mM trehalose |
| Molecular Weight (g/mol) | Predicted MW: 19.68kDa |
| Source | Chinese Hamster Ovary cell line,CHO-derived rat IFN-beta protein |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Gene Alias | Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 |
| Conjugate | Unconjugated |
| Product Line | R&D Systems |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
