Learn More
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl and PEG 3350. |
| Gene ID (Entrez) | 25712 |
| Molecular Weight (g/mol) | 15.5 kDa |
| Name | IFN-gamma |
| Quantity | 100 μg |
| Source | E. coli-derived rat IFN-gamma protein Glu23-Cys156,with an N-terminal Met |
| Storage Requirements | 3 months from date of receipt, 2 to 8° C as supplied. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
