Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat IL-17A Protein

Catalog No. 8410IL025CF
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-10I-L025CF 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-10I-L025CF Supplier R&D Systems™ Supplier No. 8410IL025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Rat IL-17A Protein is derived from CHO. The Recombinant Rat IL-17A Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_001100367
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 301289
Molecular Weight (g/mol) 15kDa
Name IL-17/IL-17A
Preparation Method Reconstitute at 100μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 13-22kDa, reducing conditions
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived rat IL-17/IL-17A protein Ala26-Ser158
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ala26 & Val27
Without Additives Carrier Protein
Structural Form Disulfide-linked homodimer
Gene Alias CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.