Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat IL-2 Protein

Catalog No. 502RL010CF
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
502RL010CF 10 μg Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol.
502RL050 50 μg Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol with BSA as a carrier protein.
502RL010 10 μg Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol with BSA as a carrier protein.
502RL050CF 50 μg Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 502RL010CF Supplier R&D Systems™ Supplier No. 502RL010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Rat IL-2 Protein is derived from E. coli. The Recombinant Rat IL-2 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol.
Gene ID (Entrez) 116562
Molecular Weight (g/mol) 16 kDa
Name IL-2
Quantity 10 μg
Source E. coli-derived rat IL-2 protein Ala21-Gln155,with and without an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.