Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat IL-23 Protein

Catalog No. 3136RL01MCF
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-36R-L01MCF 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 31-36R-L01MCF Supplier R&D Systems™ Supplier No. 3136RL01M/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Rat IL-23 Protein is derived from Sf 21 (baculovirus). The Recombinant Rat IL-23 Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number NP_072133 (p40) & NP_569094 (p19)
Formulation Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl
Gene ID (Entrez) 51561
Molecular Weight (g/mol) 58.3 kDa
Quantity 1 mg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived rat IL-23 protein, (N-terminus) Rat IL-23 p40 (Met23-Ser335) Accession &Num; NP_072133 IGSGSSRGGSGSGGSGGGGSK Rat IL-23 p19 (Leu20-Ala196) Accession &Num; NP_569094 8-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method
Biological Activity 0.1 to 0.5ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.