Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat Integrin alpha V beta 6 Protein

Catalog No. 9668AV050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
96-68A-V050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 96-68A-V050 Supplier R&D Systems™ Supplier No. 9668AV050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat Integrin alpha V beta 6 Protein is derived from CHO. The Recombinant Rat Integrin alpha V beta 6 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number XP_017456512.1 (Integrin alpha V) & Q6AYF4 (Integrin beta 6)
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 .
Gene ID (Entrez) 3685
Molecular Weight (g/mol) 115 kDa (Integrin alpha V) and 83 kDa (Integrin beta 6)
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived rat Integrin alpha V beta 6 protein, (N-terminus) Rat Integrin alpha V (Phe31-Val987) Accession &Num; XP_017456512.1 His-Pro GGGSGGGS Acidic Tail HHHHHH Rat Integrin beta 6 (Gly22-Pro708) Accession &Num; Q6AYF4 His-Pro GGGSGGGS Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Rat
Reconstitution Reconstitute at 500μg/mL in Water.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.