Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat Lymphotoxin beta R/TNFRSF3 Fc Chimera

Catalog No. 8897LR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8897LR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8897LR050 Supplier R&D Systems™ Supplier No. 8897LR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat Lymphotoxin beta R/TNFRSF3 Fc Chimera is derived from NS0. The Recombinant Rat Lymphotoxin beta R/TNFRSF3 Fc Chimera has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 4055
Molecular Weight (g/mol) Observed M.W.: 60-68kDa, reducing conditions, Theoretical M.W.: 49kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived rat Lymphotoxin beta R/TNFRSF3 protein, (N-terminus) Rat Lymphotoxin beta R (Gln29-Met222) Accession &Num; NP_001008316 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CD18, D12S370, ltbetar, LT-BETA-R, LTBR, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, LymphotoxinbR, TNF RIII, TNF Rrp, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III
Common Name Lymphotoxin beta R/TNFRSF3
Gene Symbol Ltbr
Biological Activity Measured by its ability to inhibit Lymphotoxin alpha 1/beta 2-induced IL-8 secretion in A375 human melanoma cells. Degli-Esposti, M. et al. (1997) J. Immunol. 158:1756. The ED50 for this effect is typically 0.15-0.9μg/mL.
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Rat Lymphotoxin beta R (Gln29-Met222) Accession # NP_001008316, N-terminus; IEGRMDP; Mouse IgG2a (Glu98-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.