Learn More
R&D Systems™ Recombinant Rat MANF His-tag Protein
Description
Specifications
Specifications
| Accession Number | P0C5H9.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 7873 |
| Molecular Weight (g/mol) | Observed: 18-23 kDa, under reducing conditions; Theroretical: 19 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived rat MANF protein Leu22-Leu179, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 1 mg/mL in PBS. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.