Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat Neuropilin-1 Fc Chimera Protein

Catalog No. 566N1025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
566N1025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 566N1025 Supplier R&D Systems™ Supplier No. 566N1025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Rat Neuropilin-1 Fc Chimera Protein is derived from Sf 21 (baculovirus). The Recombinant Rat Neuropilin-1 Fc Chimera Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Citrate Phosphate and NaCl.
Gene ID (Entrez) 246331
Molecular Weight (g/mol) 119 kDa (monomer)
Name Neuropilin-1 Fc Chimera
Quantity 25 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived rat Neuropilin-1 protein, (N-terminus) Rat Neuropilin-1 Phe22-Asp854 (Lys811Arg and Pro812-Gly828 del) Accession &Num; Q9QWJ9 IEGRDMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.1 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.