Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat Neuropilin-2 Fc Chimera Protein

Catalog No. 567N2025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
567N2025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 567N2025 Supplier R&D Systems™ Supplier No. 567N2025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat Neuropilin-2 Fc Chimera Protein is derived from NS0. The Recombinant Rat Neuropilin-2 Fc Chimera Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 81527
Molecular Weight (g/mol) 120 kDa (monomer)
Name Neuropilin-2 Fc Chimera
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived rat Neuropilin-2 protein, (N-terminus) Rat Neuropilin-2 Gln23-Asp857 (Val809-Asp825 del) Accession &Num; O35276 IEGRDMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.