Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Rat PBEF/Visfatin/NAMPT FLAG Protein
SDP

Catalog No. NB171698 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171698 50 μg
NB171697 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171698 Supplier Novus Biologicals™ Supplier No. NBP31814250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number Q80Z29
Concentration 0.35 mg/ml
For Use With (Application) SDS-PAGE
Formulation Liquid. 0.2um-filtered solution in PBS, pH 7.2
Gene ID (Entrez) 10135
Molecular Weight (g/mol) M.W. Theoretical: 60 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 EU/μg purified protein (LAL test; Lonza).
Gene Alias EC 2.4.2.12, MGC117256, NAmPRTase, Nampt, nicotinamide phosphoribosyltransferase, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN
Gene Symbol NAMPT
Research Category Adaptive Immunity, Cell Cycle and Replication, Immunology, Innate Immunity
Purity or Quality Grade >90% SDS-PAGE
Protein PBEF/Visfatin/NAMPT
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.