Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat PD-1 Fc Chimera Protein
SDP

Catalog No. 9814PD100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98-14P-D100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 98-14P-D100 Supplier R&D Systems™ Supplier No. 9814PD100
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Rat PD-L1/B7-H1 Fc Chimera (Catalog # 9835-B7 ) is coated at 1 μg/mL (100 μL/well), the concentration of Recombinant Rat PD-1 Fc Chimera (Catalog # 9814-PD) that produces 50% optimal binding response is typically 0.1-0.6 μg/mL.

Specifications

Accession Number NP_001100397
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and EDTA with Sucrose and Mannitol.
Gene ID (Entrez) 5133
Molecular Weight (g/mol) M.W. (Observed): 57-70 kDa, reducing conditions; M.W. (theoretical): 43 kDa
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived rat PD-1 protein Rat PD-1 (Leu25-Gln167) (N-terminus) Accession # NP_001100397 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Specific Reactivity PD-1
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL aq. soln..
Buffer Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and EDTA with Sucrose and Mannitol.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.