Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat PD-L1/B7-H1 Fc Chimera Protein

Catalog No. 9835B7050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98-35B-7050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 98-35B-7050 Supplier R&D Systems™ Supplier No. 9835B7050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. Immobilized Recombinant Rat PD-1 Fc Chimera (Catalog # 9814-PD ) at 1 μg/mL (100 μL/well) can bind Recombinant Rat PD-L1/B7-H1 Fc Chimera with an ED50 of 0.09-0.54 μg/mL.

Specifications

Accession Number NP_001178883
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 29126
Molecular Weight (g/mol) M.W. (Observed): 74-84 kDa, reducing conditions; M.W. (theoretical): 52 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived rat PD-L1/B7-H1 protein Rat B7-H1 (Phe19-Thr238) (N-terminus) Accession # NP_001178883 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species Rat
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-L1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.