Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat PD-L2/B7-DC Fc Chimera Protein
SDP

Catalog No. 9994PL100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
99-94P-L100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 99-94P-L100 Supplier R&D Systems™ Supplier No. 9994PL100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Rat PD-L2/B7-DC Fc Chimera is coated at 1 μg/mL (100 μL/well), the concentration of Recombinant Rat PD-1 Fc Chimera (Catalog # 9814-PD ) that produces 50% optimal binding response is 0.01-0.08 μg/mL

Specifications

Accession Number NP_001101052.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and EDTA with Trehalose.
Gene ID (Entrez) 80380
Molecular Weight (g/mol) M.W. (Observed): 62-73 kDa, reducing conditions; M.W. (theoretical): 49 kDa
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived rat PD-L2/B7-DC protein Rat PD-L2 (Leu20-Arg219) (N-terminus) Accession # NP_001101052.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Specific Reactivity PD-L2/B7-DC/PDCD1LG2
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL aq. soln..
Buffer Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and EDTA with Trehalose.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-L2/B7-DC/PDCD1LG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.