Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat Pentraxin 2/SAP Protein

Catalog No. 1895SA050
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 .
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
1895SA050 Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 with BSA as a carrier protein.
1895SA050CF Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 .
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 1895SA050 Supplier R&D Systems™ Supplier No. 1895SA050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat Pentraxin 2/SAP Protein is derived from NS0. The Recombinant Rat Pentraxin 2/SAP Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 with BSA as a carrier protein.
Gene ID (Entrez) 29339
Molecular Weight (g/mol) 25 kDa
Name Pentraxin 2/SAP
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived rat Pentraxin 2/SAP protein Gln21-Ser228 with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Noncovalently-linked homopentamer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.