Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Rat RBP4/Retinol-Binding Protein 4 FLAG Protein
SDP

Catalog No. NB171696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171696 50 μg
NB171695 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171696 Supplier Novus Biologicals™ Supplier No. NBP31814150UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number P04916
For Use With (Application) SDS-PAGE
Formulation Liquid. 0.2um-filtered solution in PBS, pH 7.2
Gene ID (Entrez) 5950
Molecular Weight (g/mol) M.W. Theoretical: 23 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 EU/μg purified protein (LAL test; Lonza).
Gene Alias interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma
Gene Symbol RBP4
Research Category Cancer, Diabetes Research, Lipid and Metabolism
Purity or Quality Grade >90% SDS-PAGE
Protein RBP4/Retinol-Binding Protein 4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.