Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat Serpin A12 Protein

Catalog No. 8339PI050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-39P-I050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-39P-I050 Supplier R&D Systems™ Supplier No. 8339PI050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat Serpin A12 Protein is derived from NS0. The Recombinant Rat Serpin A12 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number Q8R4Z1
Formulation Supplied as a 0.2μm filtered solution in Tris and NaCl
Gene ID (Entrez) 191570
Molecular Weight (g/mol) 47kDa
Name Serpin A12
Quality Control Testing SDS-PAGE: 45-54kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived rat Serpin A12 protein Leu20-Pro411,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Regulatory Status RUO
Termination Leu20
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5μg per lane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.