Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat sTNF RII/TNFRSF1B Protein
SDP

Catalog No. 8348R2050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-48R-2050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-48R-2050 Supplier R&D Systems™ Supplier No. 8348R2050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat sTNF RII/TNFRSF1B Protein is derived from NS0. The Recombinant Rat sTNF RII/TNFRSF1B Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q80WY6
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 156767
Molecular Weight (g/mol) 52kDa
Name TNF RII/TNFRSF1B Fc Chimera
Preparation Method Reconstitute at 500μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 62-85kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived rat TNF RII/TNFRSF1B protein, (N-terminus) Rat TNF RII/TNFRSF1B (Val23-Gly258) Accession &Num; Q80WY6 IEGRMDP Mouse IgG2 (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Val23
Structural Form Disulfide-linked Homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.