Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat VISTA/B7-H5/PD-1H Fc Chimera Protein

Catalog No. 2288B7050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-88B-7050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-88B-7050 Supplier R&D Systems™ Supplier No. 2288B7050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit anti-CD3 antibody induced IFN-gamma secretion by human peripheral blood mononuclear cells (PBMC). The ED50 for this effect is 1-6 μg/mL.

Specifications

Accession Number NP_001037765.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 64115
Molecular Weight (g/mol) M.W. (Observed): 60-70 kDa, reducing conditions; M.W. (theoretical): 44.6 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived rat VISTA/B7-H5/PD-1 H protein Rat VISTA (Met1-Ala192) (N-terminus) Accession # NP_001037765.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Specific Reactivity VISTA/B7-H5/PD-1H
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VISTA/B7-H5/PD-1 H
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.