Learn More
Novus Biologicals™ Recombinant Rhesus Macaque Flt-3 Ligand/FLT3L Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 2323 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 18.0 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of Flt-3 Ligand/FLT3L as determined by LAL method. |
| Gene Alias | FL, Flt3 ligand, FLT3L, fms-related tyrosine kinase 3 ligand, SL cytokine |
| Gene Symbol | FLT3LG |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.