Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rhesus Macaque IFN-gamma Protein

Catalog No. 961RM025CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Tris with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Tris.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
961RM025CF Lyophilized from a 0.2μm filtered solution in Tris.
961RM025 Lyophilized from a 0.2μm filtered solution in Tris with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 961RM025CF Supplier R&D Systems™ Supplier No. 961RM025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in anti-viral assays using HeLa human cervical epithelial carcinoma cells infected with encephalomyocarditis (EMC) virus. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. The ED50 for this effect is 50.0-500 ng/mL.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Tris.
Molecular Weight (g/mol) 16.9 kDa
Name IFN-gamma
Quantity 25 μg
Source E. coli-derived rhesus macaque IFN-gamma protein Gln24-Gln165,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.