Learn More
R&D Systems™ Recombinant Rhesus Macaque IL-18/IL-1F4 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT. |
| Molecular Weight (g/mol) | 18.2 kDa |
| Name | IL-18/IL-1F4 |
| Quantity | 25 μg |
| Source | E. coli-derived rhesus macaque IL-18/IL-1F4 protein Tyr37-Asp193 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | It is recommended that sterile PBS be added to the vial to prepare a working stock solution of no less than 100 μg/mL. The carrier-free protein should be used immediately upon reconstitution to avoid losses in activity due to non-specific binding to the inside surface of the vial. For long term storage as a dilute solution, a carrier protein (e.g. 0.1% HSA or BSA) should be added to the vial. |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.